missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-D-leucine, tech. 90%, Thermo Scientific Chemicals

Catalog No. p-7032835
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Catalog No. Quantity
AAH3446503 1 g
1 options

Catalog No. AAH3446503

Supplier: Thermo Scientific Chemicals H3446503

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 44978-39-4
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD08275788
InChI Key HFBHOAHFRNLZGN-MDOHGIEYNA-N
Synonym n-alpha-formyl-d-leucine, 2r-2-formamido-4-methylpentanoic acid, r-2-formamido-4-methylpentanoic acid, formyl-d-leucin, n-formyl-d-leucine, d-leucine, n-formyl, r-2-formylamino-4-methylpentanoic acid
PubChem CID 13025456
IUPAC Name (2R)-2-formamido-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC=O)C(O)=O

Specifications

Melting Point 132°C to 136°C
Quantity 1 g
Formula Weight 159.18
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Formyl-D-leucine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only