accessibility menu, dialog, popup

UserName

(R)OHOHNOH3CCH3

CAS RN 44978-39-4

IUPAC Name: (2R)-2-formamido-4-methylpentanoic acid
Synonyms: (2R)-2-formamido-4-methylpentanoic acid
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: HFBHOAHFRNLZGN-MDOHGIEYNA-N
SMILES: CC(C)C[C@@H](NC=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results