missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-S-methyl-L-cysteine, 95%

Catalog No. AAH6684214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6684214 25 g
AAH6684206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6684214 Supplier Thermo Scientific Chemicals Supplier No. H6684214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 138021-87-1 | C19H19NO4S | 357.424 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138021-87-1
Molecular Formula C19H19NO4S
Molecular Weight (g/mol) 357.424
MDL Number MFCD00237014
InChI Key SKNJDZVHMNQAGO-KRWDZBQOSA-N
Synonym fmoc-cys me-oh, fmoc-s-methyl-l-cysteine, fmoc-s-methyl-l-cyseteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylthio propanoic acid, n-alpha-9-fluorenylmethoxycarbonyl-s-methyl-l-cysteine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylsulfanyl propanoic acid, fmoc-cysteine me-oh, fmoc-l-cys me-oh, l-cysteine,n-9h-fluoren-9-ylmethoxy carbonyl-s-methyl, n-9h-fluoren-9-ylmethoxy carbonyl-s-methyl-l-cy
PubChem CID 7019705
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylsulfanylpropanoic acid
SMILES CSCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Color White
Quantity 25 g
Formula Weight 357.42
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-S-methyl-L-cysteine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only