accessibility menu, dialog, popup

UserName

CAS RN 138021-87-1

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(methylsulfanyl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(methylsulfanyl)propanoic acid
Molecular Weight (g/mol): 357.42
Molecular Formula: C19H19NO4S
InChi Key: SKNJDZVHMNQAGO-UHFFFAOYNA-N
SMILES: CSCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)