missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-S-acetamidomethyl-L-cysteine, 95%

Catalog No. p-7039838
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5930603 1 g
AAH5930606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5930603 Supplier Thermo Scientific Chemicals Supplier No. H5930603

CAS: 86060-81-3 | C21H22N2O5S | 414.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86060-81-3
Molecular Formula C21H22N2O5S
Molecular Weight (g/mol) 414.48
MDL Number MFCD00038769
InChI Key CSMYOORPUGPKAP-IBGZPJMESA-N
Synonym fmoc-cys acm-oh, fmoc-s-acetamidomethyl-l-cysteine, n alpha-fluorenylmethyloxycarbonyl-s-acetamidomethylcysteine, n, a-fmoc-s-acetaminomethyl-l-cysteine, n-fmoc-s-acetylamino methyl-l-cysteine, s-acetylamino methyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-cysteine, fmoc-cys acm, fmoc-s-acetamidomethyl-cys, l-cysteine, s-acetylamino methyl-n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 128799
IUPAC Name (2R)-3-(acetamidomethylsulfanyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(=O)NCSC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 147°C
Quantity 1 g
Beilstein 4725388
Optical Rotation −33° (c=1 in Methanol)
Formula Weight 414.47
Percent Purity 95%
Chemical Name or Material N-Fmoc-S-acetamidomethyl-L-cysteine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only