missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-N-methyl-L-threonine, 99%, Thermo Scientific Chemicals

Catalog No. AAH6317003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6317003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6317003 Supplier Thermo Scientific Chemicals Supplier No. H6317003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117106-20-4
Molecular Formula C24H29NO5
Molecular Weight (g/mol) 411.50
MDL Number MFCD02094431
InChI Key VIUVLZHFMIFLHU-VFNWGFHPSA-N
Synonym fmoc-n-me-thr tbu-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy butanoic acid, n-fmoc-n-methyl-o-tert-butyl-l-threonine, fmoc-n-methyl-o-t-butyl-l-threonine, pubchem19052, fmoc-n-me-thr but-oh, fmoc-nalpha-methyl-o-t-butyl-l-threonine, fmoc-n-alpha-methyl-o-t-butyl-l-threonine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-threonine
PubChem CID 7010372
IUPAC Name (2S,3R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](N(C)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Quantity 1 g
Formula Weight 411.49
Percent Purity 99%
Physical Form Solid
Chemical Name or Material N-Fmoc-O-tert-butyl-N-methyl-L-threonine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only