accessibility menu, dialog, popup

UserName

H3C(R)O(S)NOHOH3CCH3CH3OOH3C

CAS RN 117106-20-4

IUPAC Name: (2S,3R)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)butanoic acid
Synonyms: (2S,3R)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}butanoic acid
Molecular Weight (g/mol): 411.5
Molecular Formula: C24H29NO5
InChi Key: VIUVLZHFMIFLHU-VFNWGFHPSA-N
SMILES: C[C@@H](OC(C)(C)C)[C@H](N(C)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)