missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-O-tert-butyl-L-serine, 97%
CAS: 71989-33-8 | C22H24NO5 | 382.44 g/mol
$94.81 - $315.99
Chemical Identifiers
| CAS | 71989-33-8 |
|---|---|
| Molecular Formula | C22H24NO5 |
| Molecular Weight (g/mol) | 382.44 |
| MDL Number | MFCD00037127 |
| InChI Key | REITVGIIZHFVGU-IBGZPJMESA-M |
| Synonym | fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh |
| PubChem CID | 2724633 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
| SMILES | CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5942206
|
Thermo Scientific Chemicals
H5942206 |
5 g |
N/A
|
|
|||||
|
AAH5942214
|
Thermo Scientific Chemicals
H5942214 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 71989-33-8 | |
| 382.44 | |
| REITVGIIZHFVGU-IBGZPJMESA-M | |
| 2724633 | |
| CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O |
| C22H24NO5 | |
| MFCD00037127 | |
| fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
Specifications
| 71989-33-8 | |
| C22H24NO5 | |
| 5 g | |
| fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh | |
| +26° (c=1 in Ethyl Acetate) | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid | |
| 2724633 | |
| 98% |
| 127°C to 130°C | |
| MFCD00037127 | |
| 3632013 | |
| REITVGIIZHFVGU-IBGZPJMESA-M | |
| CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O | |
| 382.44 | |
| 383.44 | |
| N-Fmoc-O-tert-butyl-L-serine |
Safety and Handling
EINECSNumber : 276-260-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only