missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-L-serine, 97%

Catalog No. AAH5942214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5942214 25 g
AAH5942206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5942214 Supplier Thermo Scientific Chemicals Supplier No. H5942214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71989-33-8 | C22H24NO5 | 382.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71989-33-8
Molecular Formula C22H24NO5
Molecular Weight (g/mol) 382.44
MDL Number MFCD00037127
InChI Key REITVGIIZHFVGU-IBGZPJMESA-M
Synonym fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh
PubChem CID 2724633
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Melting Point 127°C to 130°C
Quantity 25 g
Beilstein 3632013
Optical Rotation +26° (c=1 in Ethyl Acetate)
Formula Weight 383.44
Percent Purity 98%
Chemical Name or Material N-Fmoc-O-tert-butyl-L-serine
EINECSNumber 276-260-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only