missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-alpha-FMOC-N-Trityl-L-histidine, 98%

Catalog No. ac360330250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC360330250 25 g
AC360330050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360330250 Supplier Thermo Scientific Chemicals Supplier No. 360330250
Only null left

CAS: 109425-51-6 | C40H33N3O4 | 619.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109425-51-6
Molecular Formula C40H33N3O4
Molecular Weight (g/mol) 619.72
InChI Key XXMYDXUIZKNHDT-QNGWXLTQSA-N
Synonym fmoc-his trt-oh, n-fmoc-n'-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, a-fmoc-n im-trityl-l-histidine, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, ambotzfaa1090
PubChem CID 11422193
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 133.0°C to 135.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 25 g
Specific Rotation 83.3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Residual Solvents 5% max. 2-propanol, THF
Formula Weight 619.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-FMOC-N-Trityl-L-histidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only