missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methylglycine, 98%

Catalog No. AAH6280606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6280606 5 g
AAH6280614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6280606 Supplier Thermo Scientific Chemicals Supplier No. H6280606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77128-70-2 | C18H19NO5 | 329.352 g/mol

N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.

Chemical Identifiers

CAS 77128-70-2
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.352
MDL Number MFCD00151926
InChI Key CUJSWOOWOONPRH-UHFFFAOYSA-N
Synonym fmoc-sarcosine monohydrate, fmoc-sarcosine hydrate, 2-9h-fluoren-9-yl methoxy carbonyl methyl amino acetic acid hydrate, 9h-fluoren-9-ylmethoxy carbonyl methyl amino acetic acid hydrate, 2-9h-fluoren-9-ylmethoxycarbonyl methyl amino acetic acid;hydrate, fmoc-sarcosinehydrate, ksc915q4d, n-9h-fluoren-9-yl methoxy carbonyl-n-methylglycine-water 1/1
PubChem CID 56763849
IUPAC Name 2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]acetic acid;hydrate
SMILES CN(CC(=O)O)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O

Specifications

Quantity 5 g
Beilstein 6660958
Solubility Information Soluble in DMF.
Formula Weight 311.33
Percent Purity 98%
Chemical Name or Material N-Fmoc-N-methylglycine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only