missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-phenylalanine, 95%

CAS: 77128-73-5 | C25H23NO4 | 401.462 g/mol

$110.33 - $411.32

Chemical Identifiers

CAS 77128-73-5
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00151938
InChI Key GBROUWPNYVBLFO-QHCPKHFHSA-N
Synonym fmoc-n-me-phe-oh, fmoc-n-methyl-l-phenylalanine, fmoc-mephe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-phenylpropanoic acid, n-fmoc-n-methyl-l-phenylalanine, l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-phenylpropanoic acid, fmoc-l-mephe-oh, ambotzfaa1403
PubChem CID 978356
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylpropanoic acid
SMILES CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6656503
View Documents
Thermo Scientific Chemicals
H6656503
1 g
Each for $110.33
Only null left
 
AAH6656506
View Documents
Thermo Scientific Chemicals
H6656506
5 g
Each for $411.32
Only null left
 
Description

Description

N-Fmoc-N-methyl-L-phenylalanine is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-N-methyl-L-phenylalanine is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in DMF. (0.3gram in 2ml)

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
Specifications

Chemical Identifiers

77128-73-5
401.462
GBROUWPNYVBLFO-QHCPKHFHSA-N
978356
CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C25H23NO4
MFCD00151938
fmoc-n-me-phe-oh, fmoc-n-methyl-l-phenylalanine, fmoc-mephe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-phenylpropanoic acid, n-fmoc-n-methyl-l-phenylalanine, l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-phenylpropanoic acid, fmoc-l-mephe-oh, ambotzfaa1403
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylpropanoic acid

Specifications

77128-73-5
C25H23NO4
4768138
Soluble in DMF. (0.3gram in 2ml)
CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
1 g
978356
95%
N-Fmoc-N-methyl-L-phenylalanine
White
MFCD00151938
fmoc-n-me-phe-oh, fmoc-n-methyl-l-phenylalanine, fmoc-mephe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-phenylpropanoic acid, n-fmoc-n-methyl-l-phenylalanine, l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-phenylpropanoic acid, fmoc-l-mephe-oh, ambotzfaa1403
GBROUWPNYVBLFO-QHCPKHFHSA-N
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylpropanoic acid
401.462
401.46
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only