missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-alanine, 95%

Catalog No. p-7054957
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6341106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6341106 Supplier Thermo Scientific Chemicals Supplier No. H6341106

CAS: 84000-07-7 | C19H19NO4 | 325.36 g/mol

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 84000-07-7
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.36
MDL Number MFCD00153384
InChI Key JOFHWKQIQLPZTC-UHFFFAOYNA-N
Synonym fmoc-n-me-ala-oh, fmoc-n-methyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino propanoic acid, fmoc-nalpha-methyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-alanine, fmoc-l-meala-oh, ambotzfaa1395
PubChem CID 688634
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]propanoic acid
SMILES CC(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 140°C
Beilstein 4329998
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 325.36
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Fmoc-N-methyl-L-alanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only