missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-N-methyl-D-leucine, 97%
CAS: 103478-63-3 | C22H25NO4 | 367.45 g/mol
$187.12 - $1366.34
Chemical Identifiers
| CAS | 103478-63-3 |
|---|---|
| Molecular Formula | C22H25NO4 |
| Molecular Weight (g/mol) | 367.45 |
| MDL Number | MFCD00235877 |
| InChI Key | BUJQSIPFDWLNDC-GNLPSFAGNA-N |
| Synonym | fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl |
| PubChem CID | 7019520 |
| IUPAC Name | (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid |
| SMILES | CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6389503
|
Thermo Scientific Chemicals
H6389503 |
1 g |
Each for $187.12
|
|
|||||
|
AAH6389506
|
Thermo Scientific Chemicals
H6389506 |
5 g |
Each for $480.16
|
|
|||||
|
AAH6389514
|
Thermo Scientific Chemicals
H6389514 |
25 g |
Each for $1,366.34
|
|
|||||
Description
Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.
Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Chemical Identifiers
| 103478-63-3 | |
| 367.45 | |
| BUJQSIPFDWLNDC-GNLPSFAGNA-N | |
| 7019520 | |
| CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C22H25NO4 | |
| MFCD00235877 | |
| fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl | |
| (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid |
Specifications
| 103478-63-3 | |
| C22H25NO4 | |
| 7350417 | |
| Soluble in clear solution (0.3 gram in 2 ml DMF). | |
| CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 1 g | |
| 7019520 | |
| 97% | |
| N-Fmoc-N-methyl-D-leucine |
| White | |
| MFCD00235877 | |
| fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl | |
| BUJQSIPFDWLNDC-GNLPSFAGNA-N | |
| (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid | |
| 367.45 | |
| 367.44 | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only