missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-D-leucine, 97%

CAS: 103478-63-3 | C22H25NO4 | 367.45 g/mol

Supplier:  Thermo Scientific Chemicals H6389506

 View more versions of this product

Catalog No. AAH6389506


Only null left
Explore available promotions

Description

Description

Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Specifications

N-Fmoc-N-methyl-D-leucine
White
MFCD00235877
fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl
BUJQSIPFDWLNDC-GNLPSFAGNA-N
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
367.45
367.44
Powder
103478-63-3
C22H25NO4
7350417
Soluble in clear solution (0.3 gram in 2 ml DMF).
CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
5 g
7019520
97%

Chemical Identifiers

103478-63-3
367.45
BUJQSIPFDWLNDC-GNLPSFAGNA-N
7019520
CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C22H25NO4
MFCD00235877
fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only