missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-valine N-succinimidyl ester, 95%
Description
N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Color | White |
| Solubility Information | Slightly soluble in water. |
| Quantity | 5 g |
| Formula Weight | 436.46 |
| Percent Purity | 95% |
| Physical Form | Powder |
| Chemical Name or Material | N-Fmoc-L-valine N-succinimidyl ester |
Safety and Handling
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only