accessibility menu, dialog, popup

UserName

H3CCH3(S)OONOOHNOO

CAS RN 130878-68-1

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methylbutanoate|FMOC-VAL-OSU
Molecular Weight (g/mol): 436.46
Molecular Formula: C24H24N2O6
InChi Key: JPJMNCROLRPFHI-QFIPXVFZSA-N
SMILES: CC(C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)