missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-valine N-succinimidyl ester, 95%

Catalog No. AAH6694606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6694606 5 g
AAH6694614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6694606 Supplier Thermo Scientific Chemicals Supplier No. H6694606

CAS: 130878-68-1 | C24H24N2O6 | 436.46 g/mol

N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 130878-68-1
Molecular Formula C24H24N2O6
Molecular Weight (g/mol) 436.46
MDL Number MFCD00153370
InChI Key JPJMNCROLRPFHI-QFIPXVFZSA-N
Synonym fmoc-val-osu, s-2,5-dioxopyrrolidin-1-yl 2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoate, n-fmoc-l-valine n-succinimidyl ester, fmoc-l-valine-osu, 2,5-dioxopyrrolidin-1-yl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylbutanoate, fmoc-l-val-osu, ambotzfaa6560, fmoc-l-valine n-hydroxysuccinimide ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-valine succinimidyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-valine succinimidyl ester
PubChem CID 11339559
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoate
SMILES CC(C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)ON1C(=O)CCC1=O

Specifications

Color White
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 436.46
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-valine N-succinimidyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only