missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-pipecolic acid, 95%

Catalog No. AAH51981MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51981MD 250 mg
AAH5198103 1 g
AAH5198106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH51981MD Supplier Thermo Scientific Chemicals Supplier No. H51981MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86069-86-5 | C21H21NO4 | 351.402 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86069-86-5
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD00235898
InChI Key CKLAZLINARHOTG-IBGZPJMESA-N
Synonym fmoc-l-homoproline, fmoc-l-pipecolic acid, fmoc-pip-oh, fmoc-hopro-oh, l-n-fmoc-pipecolic acid, fmoc-l-pip-oh, fmoc-pip, fmoc-l-piperidine-2-carboxylic acid, fmoc-s---piperidine-2-carboxylic acid, l-1-n-fmoc-pipecolinic acid
PubChem CID 983042
IUPAC Name (2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-2-carboxylic acid
SMILES C1CCN(C(C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 153°C to 158°C
Quantity 250 mg
Beilstein 4718405
Formula Weight 351.4
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-pipecolic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only