missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-norvaline, 98%

Catalog No. AAH5629003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5629003 1 g
AAH5629006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5629003 Supplier Thermo Scientific Chemicals Supplier No. H5629003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 135112-28-6 | C20H21NO4 | 339.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135112-28-6
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00155631
InChI Key JBIJSEUVWWLFGV-SFHVURJKSA-N
Synonym fmoc-nva-oh, fmoc-l-norvaline, n-9-fluorenylmethyloxycarbonyl-l-norvaline, n-fmoc-l-norvaline, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl, fmoc-norvaline, fmoc-l-nva-oh, ambotzfaa1415, pubchem14961
PubChem CID 7016885
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
SMILES CCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 151°C to 155°C
Quantity 1 g
Beilstein 5883879
Optical Rotation −21° (c=2 in DMF)
Formula Weight 339.39
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-norvaline
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only