missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-leucine, 98%

Catalog No. AAB2104006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2104006 5 g
AAB2104014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2104006 Supplier Thermo Scientific Chemicals Supplier No. B2104006

CAS: 35661-60-0 | C21H23NO4 | 353.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35661-60-0
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037133
InChI Key CBPJQFCAFFNICX-TWYLJJHKNA-N
Synonym fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
PubChem CID 1549133
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 152°C to 155°C
Quantity 5 g
Beilstein 2178254
Optical Rotation −25° (c=1 in DMF)
Formula Weight 353.42
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-leucine
EINECSNumber 252-662-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only