missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glycylglycine, 97%

Catalog No. AAH6692303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6692303 1 g
AAH6692306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6692303 Supplier Thermo Scientific Chemicals Supplier No. H6692303
Only null left

CAS: 35665-38-4 | C19H18N2O5 | 354.362 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35665-38-4
Molecular Formula C19H18N2O5
Molecular Weight (g/mol) 354.362
MDL Number MFCD00190880
InChI Key FBKUOPULLUJMOC-UHFFFAOYSA-N
Synonym fmoc-gly-gly-oh, fmoc-gly-gly, 2-2-9h-fluoren-9-yl methoxy carbonyl amino acetamido acetic acid, n-fmoc-glycylglycine, fmoc-glycyl-glycine, n-9-fluorenylmethoxycarbonylglycylglycine, glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycyl, 2-9h-fluoren-9-ylmethoxy carbonyl amino acetamido acetic acid, fmoc-glycylglycine, 2-2-9h-fluoren-9-ylmethoxycarbonylamino acetyl amino acetic acid
PubChem CID 7019069
IUPAC Name 2-[[2-(9H-fluoren-9-ylmethoxycarbonylamino)acetyl]amino]acetic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCC(=O)NCC(=O)O

Specifications

Color White
Linear Formula FmocNHCH2C(=O)NHCH2CO2H
Beilstein 2228504
Quantity 1 g
Formula Weight 354.36
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glycylglycine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only