accessibility menu, dialog, popup

UserName

CAS RN 35665-38-4

OOHNONHOOH

CAS RN 35665-38-4

IUPAC Name: 2-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetamido]acetic acid
Synonyms: (2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetamido)acetic acid
Molecular Weight (g/mol): 354.36
Molecular Formula: C19H18N2O5
InChi Key: FBKUOPULLUJMOC-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)