missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester, 98%

CAS: 86061-04-3 | C30H26F5NO6 | 591.531 g/mol

$173.93 - $764.27

Chemical Identifiers

CAS 86061-04-3
Molecular Formula C30H26F5NO6
Molecular Weight (g/mol) 591.531
MDL Number MFCD00065647
InChI Key AIDYQYOPUBOMTR-FQEVSTJZSA-N
Synonym fmoc-glu otbu-opfp, 5-tert-butyl 1-pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, 5-tert-butyl1-pentafluorophenyln-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-pentafluorophenyl 5-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4s-4-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioate, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-t-butyl ester pentafluorphenyl ester
PubChem CID 11227121
IUPAC Name 5-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioate
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6600806
View Documents
Thermo Scientific Chemicals
H6600806
5 g
Each for $173.93
Only null left
 
AAH6600814
View Documents
Thermo Scientific Chemicals
H6600814
25 g
Each for $764.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86061-04-3
591.531
AIDYQYOPUBOMTR-FQEVSTJZSA-N
11227121
CC(C)(C)OC(=O)CCC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C30H26F5NO6
MFCD00065647
fmoc-glu otbu-opfp, 5-tert-butyl 1-pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, 5-tert-butyl1-pentafluorophenyln-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-pentafluorophenyl 5-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4s-4-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioate, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-t-butyl ester pentafluorphenyl ester
5-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioate

Specifications

86061-04-3
MFCD00065647
fmoc-glu otbu-opfp, 5-tert-butyl 1-pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, 5-tert-butyl1-pentafluorophenyln-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-pentafluorophenyl 5-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4s-4-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioate, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-t-butyl ester pentafluorphenyl ester
CC(C)(C)OC(=O)CCC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
5 g
11227121
98%
N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester
C30H26F5NO6
3642680
AIDYQYOPUBOMTR-FQEVSTJZSA-N
5-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioate
591.531
591.53
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only