accessibility menu, dialog, popup

UserName

CAS RN 86061-04-3

H3CH3CCH3OOOOFFFFFNHOO

CAS RN 86061-04-3

IUPAC Name: 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanedioate
Synonyms: 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanedioate
Molecular Weight (g/mol): 591.53
Molecular Formula: C30H26F5NO6
InChi Key: AIDYQYOPUBOMTR-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)