missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals

Catalog No. p-7034369
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5205203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5205203 Supplier Thermo Scientific Chemicals Supplier No. H5205203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 193954-28-8 | Molecular Formula: C25H23NO4 | Molecular Weight: 401.462 g/mol

N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 193954-28-8
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD01863055
InChI Key DQNUGHJJKNFCND-SFHVURJKSA-N
Synonym fmoc-l-beta-homophenylalanine, fmoc-b-hophe-oh, fmoc-beta-hophe-oh, fmoc-beta-homophe-oh, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid, ambotzfaa1657, fmoc-, a-hophe-oh, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-4-phenylbutanoic acid, n-fmoc-l-beta-homophenylalanine, fmoc-s-3-amino-4-phenylbutyric acid
PubChem CID 2761539
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CC(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Beilstein 7726557
Solubility Information Slightly soluble in water.
Formula Weight 401.46
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homophenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only