missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homoleucine, 95%

CAS: 193887-44-4 | C22H25NO4 | 367.45 g/mol

$231.26 - $681.41

Chemical Identifiers

CAS 193887-44-4
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD01863059
InChI Key YLVSABQQLLRFIJ-HNNXBMFYSA-N
Synonym fmoc-beta-holeu-oh, fmoc-holeu-oh fmoc-homoleucine, fmoc-l-, fmoc-l-, a-homo-leu-oh, s-3-fmoc-amino-5-methylhexanoic acid, s-3-fmoc-amino-5-methylhexanoic acid, fmoc-?-holeu-oh, fmoc-?-holeu-oh, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-5-methylhexanoic acid, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-5-methylhexanoic acid
PubChem CID 2761527
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid
SMILES CC(C)C[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51976MD
View Documents
Thermo Scientific Chemicals
H51976MD
250 mg
Each for $231.26
Only null left
 
AAH5197603
View Documents
Thermo Scientific Chemicals
H5197603
1 g
Each for $681.41
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

193887-44-4
367.45
YLVSABQQLLRFIJ-HNNXBMFYSA-N
2761527
CC(C)C[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C22H25NO4
MFCD01863059
fmoc-beta-holeu-oh, fmoc-holeu-oh fmoc-homoleucine, fmoc-l-, fmoc-l-, a-homo-leu-oh, s-3-fmoc-amino-5-methylhexanoic acid, s-3-fmoc-amino-5-methylhexanoic acid, fmoc-?-holeu-oh, fmoc-?-holeu-oh, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-5-methylhexanoic acid, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-5-methylhexanoic acid
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid

Specifications

193887-44-4
MFCD01863059
7723942
YLVSABQQLLRFIJ-HNNXBMFYSA-N
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid
2761527
95%
C22H25NO4
250 mg
fmoc-beta-holeu-oh, fmoc-holeu-oh fmoc-homoleucine, fmoc-l-beta-homoleucine, fmoc-l-, fmoc-beta-homoleu-oh, fmoc-beta-homoleu-oh, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-5-methylhexanoic acid, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-5-methylhexanoic acid, fmoc-?-holeu-oh, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-5-methylhexanoic acid, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-5-methylhexanoic acid
CC(C)C[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
367.45
367.44
N-Fmoc-L-beta-homoleucine
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only