missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%, Thermo Scientific Chemicals

$367.18 - $367.18

Chemical Identifiers

CAS 86061-01-0
Molecular Formula C29H24F5NO6
Molecular Weight (g/mol) 577.504
MDL Number MFCD00037633
InChI Key DWYWJUBBXKYAMY-IBGZPJMESA-N
Synonym fmoc-asp otbu-opfp, n-fmoc-beta-tert-butyl-l-aspartic acid pentafluorophenyl ester, n-fmoc-l-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, n-, a-fmoc-l-aspartic acid, a-tert-butyl ester pentafluorophenyl ester, n-alpha-fmoc-l-aspartic acid beta-tert-butyl ester pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-aspartic acid 1-pentafluorophenyl 4-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino butanedioate, n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-t-butyl ester pentafluorphenyl ester
PubChem CID 11548733
IUPAC Name 4-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioate
SMILES CC(C)(C)OC(=O)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6698206
View Documents
Thermo Scientific Chemicals
H6698206
5 g
Each for $367.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86061-01-0
577.504
DWYWJUBBXKYAMY-IBGZPJMESA-N
11548733
CC(C)(C)OC(=O)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C29H24F5NO6
MFCD00037633
fmoc-asp otbu-opfp, n-fmoc-beta-tert-butyl-l-aspartic acid pentafluorophenyl ester, n-fmoc-l-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, n-, a-fmoc-l-aspartic acid, a-tert-butyl ester pentafluorophenyl ester, n-alpha-fmoc-l-aspartic acid beta-tert-butyl ester pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-aspartic acid 1-pentafluorophenyl 4-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino butanedioate, n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-t-butyl ester pentafluorphenyl ester
4-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioate

Specifications

86061-01-0
C29H24F5NO6
4222987
DWYWJUBBXKYAMY-IBGZPJMESA-N
4-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioate
577.504
577.5
Powder
White
MFCD00037633
fmoc-asp otbu-opfp, n-fmoc-beta-tert-butyl-l-aspartic acid pentafluorophenyl ester, n-fmoc-l-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, n-, a-fmoc-l-aspartic acid, a-tert-butyl ester pentafluorophenyl ester, n-alpha-fmoc-l-aspartic acid beta-tert-butyl ester pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-aspartic acid 1-pentafluorophenyl 4-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino butanedioate, n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-t-butyl ester pentafluorphenyl ester
CC(C)(C)OC(=O)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
5 g
11548733
97%
N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only