missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6698206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester | |
| White | |
| MFCD00037633 | |
| fmoc-asp otbu-opfp, n-fmoc-beta-tert-butyl-l-aspartic acid pentafluorophenyl ester, n-fmoc-l-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, n-, a-fmoc-l-aspartic acid, a-tert-butyl ester pentafluorophenyl ester, n-alpha-fmoc-l-aspartic acid beta-tert-butyl ester pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-aspartic acid 1-pentafluorophenyl 4-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino butanedioate, n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-t-butyl ester pentafluorphenyl ester | |
| CC(C)(C)OC(=O)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
| 5 g | |
| 11548733 | |
| 97% |
| 86061-01-0 | |
| C29H24F5NO6 | |
| 4222987 | |
| DWYWJUBBXKYAMY-IBGZPJMESA-N | |
| 4-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioate | |
| 577.504 | |
| 577.5 | |
| Powder |
Chemical Identifiers
| 86061-01-0 | |
| 577.504 | |
| DWYWJUBBXKYAMY-IBGZPJMESA-N | |
| 11548733 | |
| CC(C)(C)OC(=O)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
| C29H24F5NO6 | |
| MFCD00037633 | |
| fmoc-asp otbu-opfp, n-fmoc-beta-tert-butyl-l-aspartic acid pentafluorophenyl ester, n-fmoc-l-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, n-, a-fmoc-l-aspartic acid, a-tert-butyl ester pentafluorophenyl ester, n-alpha-fmoc-l-aspartic acid beta-tert-butyl ester pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-aspartic acid 1-pentafluorophenyl 4-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino butanedioate, n-alpha-9-fluorenylmethyloxycarbonyl-l-aspartic acid beta-t-butyl ester pentafluorphenyl ester | |
| 4-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioate |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only