missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid 4-methyl ester, 97%

Catalog No. AAH6377403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6377403 1 g
AAH6377406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6377403 Supplier Thermo Scientific Chemicals Supplier No. H6377403

CAS: 145038-53-5 | C20H19NO6 | 369.373 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145038-53-5
Molecular Formula C20H19NO6
Molecular Weight (g/mol) 369.373
MDL Number MFCD08458573
InChI Key HSOKCYGOTGVDHL-KRWDZBQOSA-N
Synonym fmoc-asp ome-oh, fmoc-l-aspartic acid 4-methyl ester, fmoc-aspartic acid 4-methyl ester, fmoc-l-asp ome-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid 4-methyl ester, n-, a-fmoc-l-aspartic acid, a-methyl ester
PubChem CID 45382185
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methoxy-4-oxobutanoic acid
SMILES COC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 1 g
Formula Weight 369.37
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-aspartic acid 4-methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only