accessibility menu, dialog, popup

UserName

H3COOOHONHOO

CAS RN 145038-53-5

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methoxy-4-oxobutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol): 369.37
Molecular Formula: C20H19NO6
InChi Key: HSOKCYGOTGVDHL-UHFFFAOYNA-N
SMILES: COC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)