missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid 1-tert-butyl ester, 95%

Catalog No. AAH6235503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6235503 1 g
AAH6235506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6235503 Supplier Thermo Scientific Chemicals Supplier No. H6235503
Only null left

CAS: 129460-09-9 | C23H25NO6 | 411.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129460-09-9
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.45
MDL Number MFCD00065631
InChI Key VZXQYACYLGRQJU-UHFFFAOYNA-N
Synonym fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu
PubChem CID 7017910
SMILES CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 3658045
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
Formula Weight 411.45
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-aspartic acid 1-tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only