accessibility menu, dialog, popup

UserName

H3CCH3H3COOOHONHOO

CAS RN 129460-09-9

IUPAC Name: 4-(tert-butoxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Synonyms: 4-(tert-butoxy)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoic acid
Molecular Weight (g/mol): 411.45
Molecular Formula: C23H25NO6
InChi Key: VZXQYACYLGRQJU-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results