missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid 1-allyl ester, 97%

Catalog No. p-7057081
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6641903 1 g
AAH6641906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6641903 Supplier Thermo Scientific Chemicals Supplier No. H6641903
Only null left

CAS: 144120-53-6 | C22H21NO6 | 395.411 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144120-53-6
Molecular Formula C22H21NO6
Molecular Weight (g/mol) 395.411
MDL Number MFCD00467715
InChI Key ZJMVIWUCCRKNHY-IBGZPJMESA-N
Synonym fmoc-asp-oall, fmoc-l-aspartic acid 1-allyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, fmocasp oh oallyl, ambotzfaa1355, fmoc-asp-allyl oh, fmoc-asp oh-oallyl, 1-allyl n-fmoc-l-aspartate, fmoc-l-aspartic acid a-allyl ester
PubChem CID 7020604
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
SMILES C=CCOC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Color White
Beilstein 5460108
Quantity 1 g
Formula Weight 395.41
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-aspartic acid 1-allyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only