missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-alanine, 95%

Catalog No. AAH5943814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5943814 25 g
AAH5943806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5943814 Supplier Thermo Scientific Chemicals Supplier No. H5943814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35661-39-3 | C18H17NO4 | 311.337 g/mol

N-Fmoc-L-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 35661-39-3
Molecular Formula C18H17NO4
Molecular Weight (g/mol) 311.337
MDL Number MFCD00037139
InChI Key QWXZOFZKSQXPDC-NSHDSACASA-N
Synonym fmoc-ala-oh, fmoc-l-alanine, fmoc-l-alpha-alanine, 9-fmoc-l-alanine, fmoc-l-ala-oh, n-fmoc-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine, n-9-fluorenylmethoxycarbonyl-l-alanine, fmoc-alanine, fmoc-l-ala
PubChem CID 6364642
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 147°C to 153°C
Quantity 25 g
Beilstein 2225975
Solubility Information Soluble in water.
Optical Rotation −18° (c=1 in DMF)
Formula Weight 311.34
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-alanine
EINECSNumber 252-660-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only