missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-valine, 98%

Catalog No. p-7040005
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5957003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5957003 Supplier Thermo Scientific Chemicals Supplier No. H5957003

CAS: 84624-17-9 | C20H21NO4 | 339.39 g/mol

It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Valine is one of the simplest amino acids - an isopropyl group as the side chain. This smaller side chain confers a fairly high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Valine is one of the simplest amino acids - an isopropyl group as the side chain. This smaller side chain confers a fairly high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

Solubility
Solubility in methanol (almost transparency). Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 84624-17-9
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00062953
InChI Key UGNIYGNGCNXHTR-GOSISDBHSA-N
Synonym fmoc-d-val-oh, fmoc-d-valine, n-fmoc-d-valine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-d-valine, n-9-fluorenylmethoxycarbonyl-d-valine, 9-fluorenylmethoxycarbonyl-d-valine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylbutanoic acid, n-9-fluorenylmethyloxycarbonyl-d-valine, 9h-fluoren-9-yl methoxy carbonyl-d-valine
PubChem CID 1549479
SMILES CC(C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 143°C to 144°C
Quantity 1 g
Beilstein 6489548
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoic acid
Formula Weight 339.39
Percent Purity 98%
Chemical Name or Material N-Fmoc-D-valine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only