missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-D-alanine, 95%
Description
N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.
Solubility
Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water.
Notes
Refrigerate at 4°C. Keep container tightly closed. Store away from oxidizing agent.
N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.
Solubility
Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water.
Notes
Refrigerate at 4°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 151°C to 157°C |
| Quantity | 5 g |
| Beilstein | 8025133 |
| Solubility Information | Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water. |
| Formula Weight | 311.34 |
| Percent Purity | 95% |
| Chemical Name or Material | N-Fmoc-D-alanine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only