missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-alanine, 95%

Catalog No. AAH6239514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6239514 25 g
AAH6239506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6239514 Supplier Thermo Scientific Chemicals Supplier No. H6239514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79990-15-1 | C18H17NO4 | 311.34 g/mol

N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.

Solubility
Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water.

Notes
Refrigerate at 4°C. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 79990-15-1
Molecular Formula C18H17NO4
Molecular Weight (g/mol) 311.34
MDL Number MFCD00062960
InChI Key QWXZOFZKSQXPDC-UHFFFAOYNA-N
Synonym fmoc-d-ala-oh, fmoc-d-alanine, n-fmoc-d-alanine, n-9-fluorenylmethoxycarbonyl-d-alanine, d-alanine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, n-9h-fluoren-9ylmethoxy carbonyl-d-alanine, r-2-9h-fluoren-9-yl-methoxycarbonylamino-propionic acid
PubChem CID 2724627
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 151°C to 157°C
Quantity 25 g
Beilstein 8025133
Solubility Information Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water.
Formula Weight 311.34
Percent Purity 95%
Chemical Name or Material N-Fmoc-D-alanine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only