missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-beta-alanine, 99%

CAS: 35737-10-1 | C18H16NO4 | 310.33 g/mol

$125.34 - $409.55

Chemical Identifiers

CAS 35737-10-1
Molecular Formula C18H16NO4
Molecular Weight (g/mol) 310.33
MDL Number MFCD00063328
InChI Key LINBWYYLPWJQHE-UHFFFAOYSA-M
Synonym fmoc-beta-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-beta-alanine, fmoc-beta-ala-oh, fmoc-?-ala-oh, n-fmoc-beta-alanine, fmoc-b-alanine, fmoc-b-alanine, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, a-ala-oh, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid
PubChem CID 2724630
IUPAC Name 3-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES [O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5988706
View Documents
Thermo Scientific Chemicals
H5988706
5 g
Each for $125.34
Only null left
 
AAH5988714
View Documents
Thermo Scientific Chemicals
H5988714
25 g
Each for $409.55
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water and 1% acetic acid.

Notes
Desiccate at -20°C. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

35737-10-1
310.33
LINBWYYLPWJQHE-UHFFFAOYSA-M
2724630
[O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C18H16NO4
MFCD00063328
fmoc-beta-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-beta-alanine, fmoc-beta-ala-oh, fmoc-?-ala-oh, n-fmoc-beta-alanine, fmoc-b-alanine, fmoc-b-alanine, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, a-ala-oh, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid
3-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid

Specifications

35737-10-1
C18H16NO4
MFCD00063328
2302327
Soluble in water and 1% acetic acid.
[O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
310.33
311.34
N-Fmoc-beta-alanine
142°C to 147°C
FmocNHCH2CH2CO2H
5 g
fmoc-beta-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-beta-alanine, fmoc-b-ala-oh, fmoc-b-ala-oh, n-fmoc-beta-alanine, fmoc-b-alanine, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, a-ala-oh, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid
LINBWYYLPWJQHE-UHFFFAOYSA-M
3-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
2724630
99%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only