missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-methyl-L-phenylalanine, 95%

Catalog No. AAH5207303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5207303 1 g
AAH5207306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5207303 Supplier Thermo Scientific Chemicals Supplier No. H5207303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 199006-54-7 | C25H23NO4 | 401.462 g/mol

N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Soluble in Dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 199006-54-7
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00270197
InChI Key UXLHLZHGQPDMJQ-QHCPKHFHSA-N
Synonym fmoc-phe 4-me-oh, fmoc-l-4-methylphe, fmoc-4-methyl-l-phenylalanine, fmoc-l-4-methylphenylalanine, fmoc-4-methy-l-phenylalanine, fmoc-l-phe 4-me-oh, fmoc-p-me-phe-oh, n-fmoc-4-methyl-l-phenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-p-tolyl propanoic acid, phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-methyl
PubChem CID 7129826
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Beilstein 8799411
Solubility Information Soluble in Dimethylformamide(DMF).
Formula Weight 401.46
Percent Purity 95%
Chemical Name or Material N-Fmoc-4-methyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only