accessibility menu, dialog, popup

UserName

CH3(S)HNOOOHO

CAS RN 199006-54-7

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methylphenyl)propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid
Molecular Weight (g/mol): 401.46
Molecular Formula: C25H23NO4
InChi Key: UXLHLZHGQPDMJQ-QHCPKHFHSA-N
SMILES: CC1=CC=C(C[C@H](NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)