missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-4-iodo-D-phenylalanine, 98%
CAS: 205526-29-0 | C24H20INO4 | 513.33 g/mol
$111.04 - $441.33
Chemical Identifiers
| CAS | 205526-29-0 |
|---|---|
| Molecular Formula | C24H20INO4 |
| Molecular Weight (g/mol) | 513.33 |
| MDL Number | MFCD00672557 |
| InChI Key | LXOXXTQKKRJNNB-UHFFFAOYNA-N |
| Synonym | fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc |
| PubChem CID | 16213204 |
| SMILES | OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6329203
|
Thermo Scientific Chemicals
H6329203 |
1 g |
Each for $111.04
|
|
|||||
|
AAH6329206
|
Thermo Scientific Chemicals
H6329206 |
5 g |
Each for $441.33
|
|
|||||
Description
N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Sparingly Soluble in water (6.7E-4 g/L) (25°C).
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
| 205526-29-0 | |
| 513.33 | |
| LXOXXTQKKRJNNB-UHFFFAOYNA-N | |
| 16213204 |
| C24H20INO4 | |
| MFCD00672557 | |
| fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc | |
| OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
| 205526-29-0 | |
| MFCD00672557 | |
| fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc | |
| LXOXXTQKKRJNNB-UHFFFAOYNA-N | |
| 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-iodophenyl)propanoic acid | |
| 513.33 | |
| 513.32 | |
| 98% | |
| N-Fmoc-4-iodo-D-phenylalanine |
| C24H20INO4 | |
| 9159551 | |
| Sparingly Soluble in water (6.7E-4g/L) (25°C). | |
| OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 1 g | |
| 16213204 | |
| Light sensitive | |
| Solid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only