missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-iodo-D-phenylalanine, 98%

CAS: 205526-29-0 | C24H20INO4 | 513.33 g/mol

Supplier:  Thermo Scientific Chemicals H6329203

 View more versions of this product

Catalog No. AAH6329203


Only null left
Explore available promotions

Description

Description

N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly Soluble in water (6.7E-4 g/L) (25°C).

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

N-Fmoc-4-iodo-D-phenylalanine
C24H20INO4
9159551
Sparingly Soluble in water (6.7E-4g/L) (25°C).
OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1 g
16213204
Light sensitive
Solid
205526-29-0
MFCD00672557
fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc
LXOXXTQKKRJNNB-UHFFFAOYNA-N
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-iodophenyl)propanoic acid
513.33
513.32
98%

Chemical Identifiers

205526-29-0
513.33
LXOXXTQKKRJNNB-UHFFFAOYNA-N
16213204
C24H20INO4
MFCD00672557
fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc
OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only