missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(4-thiazolyl)-L-alanine, 95%

Catalog No. p-7034388
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52071MD 250 mg
AAH5207103 1 g
AAH5207106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52071MD Supplier Thermo Scientific Chemicals Supplier No. H52071MD
Only null left

CAS: 205528-32-1 | C21H18N2O4S | 394.445 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 205528-32-1
Molecular Formula C21H18N2O4S
Molecular Weight (g/mol) 394.445
MDL Number MFCD00672568
InChI Key LSBAZFASKHLHKB-IBGZPJMESA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiazol-4-yl propanoic acid, fmoc-l-4-thiazolyl-alanine, fmoc-beta-4-thiazolyl-ala-oh, n-fmoc-3-4-thiazolyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1,3-thiazol-4-yl propanoic acid, fmoc-3-4-thiazoyl-l-alanine, fmoc-4-thi-oh, fmoc-3-4-thiazolyl-alanine, fmoc-, a-4-thiazolyl-ala-oh, fmoc-3-4-thiazolyl-l-alanine
PubChem CID 7020858
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1,3-thiazol-4-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=N4)C(=O)O

Specifications

Melting Point 178°C to 179°C
Quantity 250 mg
Formula Weight 394.44
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(4-thiazolyl)-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only