missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3,3-diphenyl-L-alanine, 95%

Catalog No. p-7034490
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5217603 1 g
AAH5217606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5217603 Supplier Thermo Scientific Chemicals Supplier No. H5217603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 201484-50-6 | C30H25NO4 | 463.533 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 201484-50-6
Molecular Formula C30H25NO4
Molecular Weight (g/mol) 463.533
MDL Number MFCD00672337
InChI Key PENQOTJCVODUQU-NDEPHWFRSA-N
Synonym fmoc-l-3,3-diphenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3,3-diphenylpropanoic acid, fmoc-3,3-diphenyl-l-alanine, fmoc-beta-phenyl-phe-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3,3-diphenyl-propanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3,3-diphenylpropanoic acid, fmoc-3,3-diphenyl-d-alaine, fmoc-3,3-diphenyl-l-alaine, fmoc-,-diphenyl-ala-oh, fmoc-ala 3,3-diphenyl-oh
PubChem CID 2761488
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3,3-diphenylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35

Specifications

Melting Point 125°C to 129°C
Quantity 1 g
UN Number UN3077
Beilstein 8873562
Formula Weight 463.53
Percent Purity 95%
Chemical Name or Material N-Fmoc-3,3-diphenyl-L-alanine
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only