missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(2-naphthyl)-D-alanine, 95%

Catalog No. AAH5197403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197403 1 g
AAH5197406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5197403 Supplier Thermo Scientific Chemicals Supplier No. H5197403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 138774-94-4 | C28H23NO4 | 437.50 g/mol

N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 138774-94-4
Molecular Formula C28H23NO4
Molecular Weight (g/mol) 437.50
MDL Number MFCD00077044
InChI Key JYUTZJVERLGMQZ-AREMUKBSSA-N
Synonym fmoc-d-2-nal-oh, fmoc-3-2-naphthyl-d-alanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, fmoc-d-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-d-alanine, fmoc-d-2-naphthylalanine, s-n-fmoc-2-naphthylalanine, fmoc-beta-2-naphthyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, pubchem11948
PubChem CID 7130590
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
SMILES OC(=O)[C@@H](CC1=CC2=CC=CC=C2C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 7864489
Solubility Information Slightly soluble in water.
Formula Weight 437.49
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(2-naphthyl)-D-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only