missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(1-naphthyl)-L-alanine, 98%

Catalog No. AAH5196703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5196703 1 g
AAH5196706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5196703 Supplier Thermo Scientific Chemicals Supplier No. H5196703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96402-49-2 | C28H23NO4 | 437.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96402-49-2
Molecular Formula C28H23NO4
Molecular Weight (g/mol) 437.50
MDL Number MFCD00151914
InChI Key ORWNVJDLEMVDLV-SANMLTNESA-N
Synonym fmoc-3-1-naphthyl-l-alanine, fmoc-1-nal-oh, fmoc-l-1-naphthylalanine, fmoc-beta-1-naphthyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, fmoc-3-1 naphthyl-l-alanine, fmoc-l-3-1-naphthyl-alanine, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, n-fmoc-3-1-naphthyl-l-alanine
PubChem CID 2734474
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-1-ylpropanoic acid
SMILES OC(=O)[C@H](CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 171°C to 183°C
Quantity 1 g
Beilstein 4829118
Formula Weight 437.49
Percent Purity 98%
Chemical Name or Material N-Fmoc-3-(1-naphthyl)-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only