missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-2-cyclohexyl-L-glycine, 98%

CAS: 161321-36-4 | C23H25NO4 | 379.46 g/mol

Supplier:  Thermo Scientific Chemicals H6358906

 View more versions of this product

Catalog No. AAH6358906


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Fmoc-2-cyclohexyl-L-glycine
C23H25NO4
fmoc-chg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, s-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cyclohexylglycine, ambotzfaa1713, fmoc-cyclohexyl-gly-oh, bicr147, n-fmoc-2-cyclohexyl-l-glycine, s-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylglycine
OC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1CCCCC1
5 g
6976353
98%
161321-36-4
MFCD00190878
BWQQGHPODCJZDB-UHFFFAOYNA-N
2-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid
379.46
379.45
Solid

Chemical Identifiers

161321-36-4
379.46
BWQQGHPODCJZDB-UHFFFAOYNA-N
6976353
C23H25NO4
MFCD00190878
fmoc-chg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, s-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cyclohexylglycine, ambotzfaa1713, fmoc-cyclohexyl-gly-oh, bicr147, n-fmoc-2-cyclohexyl-l-glycine, s-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylglycine
OC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1CCCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only