missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-2-cyclohexyl-L-glycine, 98%

Catalog No. p-7055096
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63589MD 250 mg
AAH6358903 1 g
AAH6358906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63589MD Supplier Thermo Scientific Chemicals Supplier No. H63589MD

CAS: 161321-36-4 | C23H25NO4 | 379.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161321-36-4
Molecular Formula C23H25NO4
Molecular Weight (g/mol) 379.46
MDL Number MFCD00190878
InChI Key BWQQGHPODCJZDB-UHFFFAOYNA-N
Synonym fmoc-chg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, s-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cyclohexylglycine, ambotzfaa1713, fmoc-cyclohexyl-gly-oh, bicr147, n-fmoc-2-cyclohexyl-l-glycine, s-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylglycine
PubChem CID 6976353
SMILES OC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1CCCCC1

Specifications

IUPAC Name 2-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid
Quantity 250 mg
Formula Weight 379.45
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-2-cyclohexyl-L-glycine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only