missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-1-trityl-D-histidine, 98%

Catalog No. AAH6250406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6250406 5 g
AAH6250414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6250406 Supplier Thermo Scientific Chemicals Supplier No. H6250406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 135610-90-1 | C40H33N3O4 | 619.721 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135610-90-1
Molecular Formula C40H33N3O4
Molecular Weight (g/mol) 619.721
MDL Number MFCD00077061
InChI Key XXMYDXUIZKNHDT-DIPNUNPCSA-N
Synonym fmoc-d-his trt-oh, n-fmoc-n'-trityl-d-histidine, d-histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, pubchem18930, fmoc-d-his 1-trt-oh, fmoc-d-his-trt-oh, n-fmoc-1-trityl-d-histidine, fmoc-n-im-trityl-d-histidine, nalpha-fmoc-nim-trityl-d-histidine
PubChem CID 44828577
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

Quantity 5 g
Optical Rotation −80° (c=1 in Chloroform)
Formula Weight 619.71
Percent Purity 98%
Chemical Name or Material N-Fmoc-1-trityl-D-histidine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only